Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030166.1 | NP_001025337.1 | 12 - 13 | 2232 | 70 | |
GenBank mRNA | AY974804.1 | - | 12 - 13 | 2232 | 70 | |
BC163641.1 | - | 12 - 13 | 2316 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |