Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map4k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040358.1 | NP_001035448.1 | 26 - 27 | 2159 | 102 | |
GenBank mRNA | BC115221.1 | - | 26 - 27 | 2159 | 102 | |
BC162619.1 | - | 26 - 27 | 2257 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |