Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045002.1 | NP_001038467.1 | 12 - 13 | 1646 | 67 | |
NM_001327842.1 | NP_001314771.1 | 13 - 14 | 2855 | 67 | ||
XM_005164650.4 | XP_005164707.1 | 12 - 13 | 2477 | 67 | ||
XM_009300161.2 | XP_009298436.1 | 12 - 13 | 1643 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |