Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045002.1 | NP_001038467.1 | 4 - 5 | 603 | 90 | |
NM_001327842.1 | NP_001314771.1 | 5 - 6 | 1815 | 90 | ||
XM_005164650.4 | XP_005164707.1 | 5 - 6 | 1539 | 90 | ||
XM_009300161.2 | XP_009298436.1 | 4 - 5 | 603 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |