Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045087.1 | NP_001038552.1 | 3 - 4 | 315 | 69 | |
GenBank mRNA | AB124553.1 | - | 3 - 4 | 352 | 69 | |
AY681353.1 | - | 3 - 4 | 315 | 69 | ||
BC076449.1 | - | 3 - 4 | 294 | 69 | ||
BC134025.1 | - | 3 - 4 | 347 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |