Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045856.2 | NP_001039321.2 | 5 - 6 | 1343 | 149 | |
XM_021469354.1 | XP_021325029.1 | 5 - 6 | 1447 | 149 | ||
GenBank mRNA | EF030485.1 | - | 5 - 6 | 1343 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Probe spans exons |