Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076745.2 | NP_001070213.2 | 3 - 4 | 750 | 75 | |
GenBank mRNA | BC124532.1 | - | 3 - 4 | 545 | 75 | |
BC164409.1 | - | 3 - 4 | 436 | 75 | ||
CB359574.1 | - | 2 - 3 | 177 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |