Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076745.2 | NP_001070213.2 | 4 - 5 | 842 | 150 | |
GenBank mRNA | BC124532.1 | - | 4 - 5 | 637 | 150 | |
BC164409.1 | - | 4 - 5 | 528 | 150 | ||
CB359574.1 | - | 3 - 4 | 269 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |