Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ecsit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242938.1 | NP_001229867.1 | 5 - 6 | 981 | 86 | |
XM_009302997.3 | XP_009301272.1 | 5 - 6 | 1005 | 86 | ||
GenBank mRNA | BC124208.1 | - | 5 - 6 | 941 | 86 | |
BC164678.1 | - | 4 - 5 | 867 | 86 | ||
DY551107.1 | - | 3 - 4 | 481 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |