Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem41ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213083.1 | NP_998248.1 | 1 - 2 | 196 | 77 | |
GenBank mRNA | BC068328.1 | - | 1 - 2 | 196 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |