Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lman2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081747.2 | NP_001075216.2 | 5 - 6 | 732 | 107 | |
XM_005173172.4 | XP_005173229.1 | 5 - 6 | 698 | 107 | ||
GenBank mRNA | BC133153.1 | - | 5 - 6 | 671 | 107 | |
EH442331.1 | - | 5 - 6 | 702 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |