Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130623.2 | NP_001124095.1 | 2 - 3 | 469 | 71 | |
GenBank mRNA | AY772712.1 | - | 2 - 3 | 385 | 71 | |
BC163074.1 | - | 2 - 3 | 469 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |