Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105594.1 | NP_001099064.1 | 3 - 4 | 1044 | 75 | |
GenBank mRNA | BC133172.1 | - | 3 - 4 | 813 | 75 | |
BC142873.1 | - | 3 - 4 | 810 | 75 | ||
BC153383.1 | - | 3 - 4 | 1044 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |