Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dync2i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105690.1 | NP_001099160.1 | 1 - 2 | 414 | 86 | |
XM_021468923.1 | XP_021324598.1 | 1 - 2 | 433 | 86 | ||
GenBank mRNA | BC095866.1 | - | 1 - 2 | 416 | 86 | |
BC152526.1 | - | 1 - 2 | 414 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |