Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ednrba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131197.1 | NP_571272.1 | 1 - 2 | 564 | 127 | |
GenBank mRNA | AF275636.1 | - | 1 - 2 | 564 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Danio rerio Probe spans exons |