Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131348.2 | NP_571423.1 | 5 - 6 | 759 | 89 | |
XM_005159278.4 | XP_005159335.1 | 6 - 7 | 1039 | 89 | ||
XM_021467581.1 | XP_021323256.1 | 6 - 7 | 1085 | 89 | ||
XM_021467582.1 | XP_021323257.1 | 6 - 7 | 1066 | 89 | ||
XM_021467583.1 | XP_021323258.1 | 6 - 7 | 1058 | 89 | ||
XM_021467584.1 | XP_021323259.1 | 5 - 6 | 864 | 89 | ||
XM_021467585.1 | XP_021323260.1 | 5 - 6 | 783 | 89 | ||
GenBank mRNA | AF177538.1 | - | 4 - 5 | 619 | 89 | |
AJ249590.1 | - | 5 - 6 | 700 | 89 | ||
BC056588.1 | - | 5 - 6 | 748 | 89 | ||
BC066571.1 | - | 5 - 6 | 759 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |