Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: raraa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131406.2 | NP_571481.2 | 4 - 5 | 1228 | 102 | |
XM_005156052.4 | XP_005156109.1 | 6 - 7 | 1212 | 102 | ||
GenBank mRNA | BC092692.1 | - | 4 - 5 | 1228 | 102 | |
KF148618.1 | - | 4 - 5 | 846 | 102 | ||
L03398.1 | - | 4 - 5 | 710 | 102 | ||
S74155.1 | - | 4 - 5 | 799 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |