Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131635.3 | NP_571710.1 | 2 - 3 | 721 | 78 | |
GenBank mRNA | AF283555.1 | - | 2 - 3 | 401 | 78 | |
BC128817.1 | - | 2 - 3 | 719 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |