Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp1a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131690.1 | NP_571765.1 | 18 - 19 | 2735 | 76 | |
GenBank mRNA | AY008375.1 | - | 18 - 19 | 2735 | 76 | |
BC085663.1 | - | 18 - 19 | 2800 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |