Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: selenot1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178290.6 | NP_840075.2 | 5 - 6 | 685 | 70 | |
GenBank mRNA | CF998986.1 | - | 5 - 6 | 650 | 70 | |
EH438162.1 | - | 5 - 6 | 683 | 70 | ||
EH473273.1 | - | 1 - 2 | 655 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Danio rerio Probe spans exons |