Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gad1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194419.1 | NP_919400.1 | 11 - 12 | 1260 | 88 | |
XM_009302103.3 | XP_009300378.1 | 11 - 12 | 1252 | 88 | ||
XM_009302104.3 | XP_009300379.1 | 10 - 11 | 1437 | 88 | ||
GenBank mRNA | AB183390.1 | - | 11 - 12 | 1444 | 88 | |
AF017266.1 | - | 5 - 6 | 88 | |||
AF042374.1 | - | 5 - 6 | 470 | 88 | ||
BC047851.1 | - | 11 - 12 | 1260 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |