Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v0b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199561.2 | NP_955855.2 | 5 - 6 | 540 | 97 | |
GenBank mRNA | BC058877.1 | - | 5 - 6 | 528 | 97 | |
EH446151.1 | - | 5 - 6 | 542 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |