Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pabpc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199839.1 | NP_956133.1 | 9 - 10 | 1339 | 66 | |
GenBank mRNA | AY398370.1 | - | 9 - 10 | 1350 | 66 | |
BC044513.1 | - | 9 - 10 | 1339 | 66 | ||
BC165470.1 | - | 8 - 9 | 1277 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |