Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199929.1 | NP_956223.1 | 11 - 12 | 1794 | 63 | |
GenBank mRNA | BC045367.1 | - | 11 - 12 | 1794 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |