Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sybl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200266.1 | NP_956560.1 | 2 - 3 | 354 | 84 | |
XM_005157135.4 | XP_005157192.1 | 2 - 3 | 84 | |||
GenBank mRNA | BC049034.1 | - | 2 - 3 | 354 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |