Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtmr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200394.1 | NP_956688.1 | 12 - 13 | 1631 | 72 | |
XM_021476036.1 | XP_021331711.1 | 7 - 8 | 777 | 72 | ||
GenBank mRNA | BC053411.1 | - | 12 - 13 | 1631 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |