Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hao2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200483.2 | NP_956777.1 | 2 - 3 | 283 | 93 | |
XM_005167544.4 | XP_005167601.1 | 3 - 4 | 359 | 93 | ||
XM_005167545.4 | XP_005167602.1 | 2 - 3 | 298 | 93 | ||
GenBank mRNA | BC055205.1 | - | 2 - 3 | 284 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |