Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hao2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200483.2 | NP_956777.1 | 6 - 7 | 1097 | 94 | |
XM_005167544.4 | XP_005167601.1 | 7 - 8 | 1173 | 94 | ||
XM_005167545.4 | XP_005167602.1 | 6 - 7 | 1112 | 94 | ||
GenBank mRNA | BC055205.1 | - | 6 - 7 | 1098 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |