Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200555.2 | NP_956849.2 | 3 - 4 | 322 | 57 | |
XM_005158980.4 | XP_005159037.1 | 3 - 4 | 271 | 57 | ||
GenBank mRNA | BC056552.1 | - | 3 - 4 | 296 | 57 | |
EH548629.1 | - | 3 - 4 | 361 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |