Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200807.1 | NP_957101.1 | 3 - 4 | 325 | 129 | |
XM_005162679.4 | XP_005162736.1 | 3 - 4 | 367 | 129 | ||
GenBank mRNA | BC059679.1 | - | 3 - 4 | 325 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |