Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200836.2 | NP_957130.2 | 5 - 6 | 627 | 86 | |
XM_017352206.2 | XP_017207695.1 | 5 - 6 | 612 | 86 | ||
GenBank mRNA | BC060935.1 | - | 5 - 6 | 537 | 86 | |
BC091655.1 | - | 5 - 6 | 632 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |