Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200836.2 | NP_957130.2 | 7 - 8 | 817 | 59 | |
XM_017352206.2 | XP_017207695.1 | 7 - 8 | 802 | 59 | ||
GenBank mRNA | BC060935.1 | - | 7 - 8 | 727 | 59 | |
BC091655.1 | - | 7 - 8 | 822 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |