Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201034.2 | NP_957328.2 | 2 - 3 | 389 | 101 | |
GenBank mRNA | BC055159.1 | - | 2 - 3 | 334 | 101 | |
CT621066.2 | - | 2 - 3 | 248 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |