Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205754.1 | NP_991317.1 | 1 - 2 | 160 | 120 | |
GenBank mRNA | AY398401.1 | - | 1 - 2 | 160 | 120 | |
BC094958.1 | - | 1 - 2 | 147 | 120 | ||
BC165057.1 | - | 1 - 2 | 147 | 120 | ||
DQ388034.1 | - | 1 - 2 | 113 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |