Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212608.1 | NP_997773.1 | 1 - 2 | 197 | 65 | |
GenBank mRNA | BC044505.1 | - | 1 - 2 | 197 | 65 | |
BC067549.1 | - | 1 - 2 | 218 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |