Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212655.2 | NP_997820.2 | 17 - 18 | 2485 | 95 | |
GenBank mRNA | BC056710.1 | - | 17 - 18 | 2461 | 95 | |
BC066556.1 | - | 17 - 18 | 2449 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |