Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ren | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212860.1 | NP_998025.1 | 8 - 9 | 1065 | 73 | |
GenBank mRNA | AY167037.1 | - | 8 - 9 | 1065 | 73 | |
BC162339.1 | - | 8 - 9 | 1065 | 73 | ||
BC162351.1 | - | 8 - 9 | 1065 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |