Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sort1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213230.1 | NP_998395.1 | 15 - 16 | 1890 | 58 | |
XM_005172902.4 | XP_005172959.1 | 14 - 15 | 1939 | 58 | ||
GenBank mRNA | BC068433.1 | - | 15 - 16 | 1890 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |