Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213245.1 | NP_998410.1 | 4 - 5 | 360 | 143 | |
XM_005160456.2 | XP_005160513.1 | 4 - 5 | 474 | 143 | ||
GenBank mRNA | BC063962.1 | - | 4 - 5 | 360 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |