Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213494.2 | NP_998659.2 | 4 - 5 | 1054 | 84 | |
GenBank mRNA | BC055234.1 | - | 4 - 5 | 651 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |