Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002530.1 | NP_001002530.1 | 3 - 4 | 491 | 67 | |
XM_005166825.4 | XP_005166882.1 | 3 - 4 | 528 | 67 | ||
GenBank mRNA | BC076367.1 | - | 3 - 4 | 491 | 67 | |
BC165042.1 | - | 2 - 3 | 374 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |