Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002530.1 | NP_001002530.1 | 4 - 5 | 615 | 113 | |
XM_005166825.4 | XP_005166882.1 | 4 - 5 | 652 | 113 | ||
GenBank mRNA | BC076367.1 | - | 4 - 5 | 615 | 113 | |
BC165042.1 | - | 3 - 4 | 498 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |