Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b4galt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003417.1 | NP_001003417.1 | 3 - 4 | 1126 | 74 | |
XM_021481002.1 | XP_021336677.1 | 4 - 5 | 1212 | 74 | ||
GenBank mRNA | AY648740.1 | - | 3 - 4 | 1126 | 74 | |
BC092934.1 | - | 3 - 4 | 1064 | 74 | ||
BC164341.1 | - | 3 - 4 | 641 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |