Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003840.1 | NP_001003840.1 | 7 - 8 | 776 | 94 | |
GenBank mRNA | AY648829.1 | - | 7 - 8 | 776 | 94 | |
BC162284.1 | - | 7 - 8 | 774 | 94 | ||
BC162298.1 | - | 7 - 8 | 774 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |