Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005981.1 | NP_001005981.1 | 6 - 7 | 817 | 100 | |
XM_005170550.4 | XP_005170607.1 | 6 - 7 | 919 | 100 | ||
GenBank mRNA | BC083429.1 | - | 6 - 7 | 817 | 100 | |
BC163998.1 | - | 6 - 7 | 800 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |