Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006024.1 | NP_001006024.1 | 6 - 7 | 913 | 95 | |
GenBank mRNA | BC083397.1 | - | 6 - 7 | 913 | 95 | |
BC164011.1 | - | 6 - 7 | 887 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |