Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zmynd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007304.1 | NP_001007305.1 | 3 - 4 | 555 | 96 | |
XM_021478243.1 | XP_021333918.1 | 3 - 4 | 626 | 96 | ||
GenBank mRNA | BC085655.1 | - | 3 - 4 | 555 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |