Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007331.1 | NP_001007332.1 | 2 - 3 | 291 | 94 | |
XM_009293191.3 | XP_009291466.1 | 2 - 3 | 506 | 94 | ||
XM_009293192.3 | XP_009291467.1 | 2 - 3 | 226 | 94 | ||
GenBank mRNA | BC085557.1 | - | 2 - 3 | 291 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |