Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007331.1 | NP_001007332.1 | 5 - 6 | 479 | 141 | |
XM_009293191.3 | XP_009291466.1 | 5 - 6 | 694 | 141 | ||
XM_009293192.3 | XP_009291467.1 | 5 - 6 | 414 | 141 | ||
GenBank mRNA | BC085557.1 | - | 5 - 6 | 479 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |