Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smyd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013550.1 | NP_001013568.1 | 2 - 3 | 375 | 90 | |
GenBank mRNA | BC091465.1 | - | 2 - 3 | 375 | 90 | |
DQ851823.1 | - | 2 - 3 | 258 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |